Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-5-nitrobenzonitrile 98%

Supplier: Ambeed
Warning

A142595-1G A142595-5G A142595-100G A142595-25G
76767-880EA 5.15 USD InStock
76767-880 76767-882 76767-878 76767-884
2-Chloro-5-nitrobenzonitrile 98%
2-Chloro-5-nitrobenzonitrile
Caution: for Research Use Only


Formula: C₇H₃ClN₂O₂
MW: 182.57 g/mol
MDL Number: MFCD00007292
CAS Number: 16588-02-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty