Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromoisophthalic acid 98%

Supplier: Ambeed
Warning

A177641-5G A177641-25G A177641-1G A177641-100G
76769-450EA 12.36 USD InStock
76769-450 76769-452 76769-454 76769-448
4-Bromoisophthalic acid 98%
4-Bromoisophthalic acid
Caution: for Research Use Only


Formula: C₈H₅BrO₄
MW: 245.03 g/mol
MDL Number: MFCD00002404
CAS Number: 6939-93-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty