Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluoro-3-nitrotoluene 98%

Supplier: Ambeed
Warning

A266957-500G A266957-5G A266957-100G A266957-25G A266957-10G
76772-038EA 17.51 USD InStock
76772-038 76772-036 76772-040 76772-034 76772-042
4-Fluoro-3-nitrotoluene 98%
4-Fluoro-3-nitrotoluene
Caution: for Research Use Only


Formula: C₇H₆FNO₂
MDL Number: MFCD00007060
CAS Number: 446-11-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty