Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Trimethylsilyl)-3-butyn-1-ol 98%

Supplier: Ambeed
Danger
Synonyms: 4-Trimethylsilyl-3-butyn-1-ol

A308264-5G
76772-744EA 26.78 USD InStock
76772-744
4-(Trimethylsilyl)-3-butyn-1-ol 98%
4-(Trimethylsilyl)-3-butyn-1-ol
Caution: for Research Use Only


Formula: C₇H₁₄OSi
MW: 142.27 g/mol
MDL Number: MFCD00190217
CAS Number: 2117-12-6
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty