Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(2-Bromophenyl)acetonitrile 98%

Supplier: Ambeed
Warning

A378382-5G A378382-500G A378382-25G A378382-1000G A378382-100G A378382-10G
76773-552EA 46.35 USD InStock
76773-552 76773-554 76773-556 76773-558 76840-970 76773-550
(2-Bromophenyl)acetonitrile 98%
(2-Bromophenyl)acetonitrile
Caution: for Research Use Only


Formula: C₈H₆BrN
MW: 196.05 g/mol
MDL Number: MFCD00001896
CAS Number: 19472-74-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty