Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Dimethylacrylamide 97% stabilized

Supplier: Ambeed
Danger

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A514826-100G A514826-25G
76774-600EA 6.18 USD InStock
76774-600 76774-598
N,N-Dimethylacrylamide 97% stabilized
N,N-Dimethylacrylamide
Caution: for Research Use Only


Formula: C₅H₉NO
MDL Number: MFCD00008626
CAS Number: 2680-03-7
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty