Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluorophenyl isocyanate 97%

Supplier: Ambeed
Danger
Synonyms: 4-Fluorophenylisocyanate1-Fluoro-4-isocyanatobenzene

A655096-5G
76775-582EA 10.3 USD InStock
76775-582
4-Fluorophenyl isocyanate 97%
4-Fluorophenyl isocyanate
Caution: for Research Use Only


Formula: C₇H₄FNO
MW: 137.11 g/mol
MDL Number: MFCD00002023
CAS Number: 1195-45-5
UN: 2478
ADR: 3,6.1,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty