Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Hexenoic acid 95%

Supplier: Ambeed
Danger
Synonyms: 5-Hexenoicacid

A675098-250MG A675098-5G A675098-1G
76775-712EA 64.89 USD InStock
76775-712 76775-710 76775-708
5-Hexenoic acid 95%
5-Hexenoic acid
Caution: for Research Use Only


Formula: C₆H₁₀O₂
MW: 114.14 g/mol
MDL Number: MFCD00046558
CAS Number: 1577-22-6
UN: 3265
ADR: 8,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty