Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Bromovaleryl chloride 97%

Supplier: Ambeed
Danger
Synonyms: 5-Bromovalerylchloride5-Bromopentanoyl chloride

A756630-5G A756630-1G A756630-25G
76776-242EA 8.24 USD InStock
76776-242 76776-246 76776-244
5-Bromovaleryl chloride 97%
5-Bromovaleryl chloride
Caution: for Research Use Only


Formula: BrH₂C(CH₂)₃COOCl
MW: 199.47 g/mol
MDL Number: MFCD00013660
CAS Number: 4509-90-4
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty