Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chloro-1,10-phenanthroline 98%

Supplier: Ambeed
Danger

A933369-1G A933369-5G
76777-002EA 53.56 USD InStock
76777-002 76777-004
5-Chloro-1,10-phenanthroline 98%
5-Chloro-1,10-phenanthroline
Caution: for Research Use Only


Formula: C₁₂H₇ClN₂
MDL Number: MFCD00004980
CAS Number: 4199-89-7
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty