Would you like to log in again?
Parker MegaBond™ Nominal Melt Blown Filter Cartridges
Supplier: Parker Hannifin
Parker
MBN5M10
76249-994CS 253.74 USD InStock
76249-994
Parker MegaBond™ Nominal Melt Blown Filter Cartridges
Filters Filter Cartridges
MegaBond™ Nominal melt blown filter cartridges are the most economical high purity filter cartridges available. Featuring a graded density matrix of uniform polypropylene fibers, the filter provides consistent filtration for a variety of fluids. No fiber finish or surfactants are present to generate extractables leading to foaming or other undesirable effects on the filtrate.
  • Thermally bonded melt blown fiber matrix provides dimensionally stable construction
  • Continuous fiber matrix prevents media migration and ensures consistent quality filtration performance
  • Finish-free construction provides optimum fluid purity and eliminates foaming condition
  • Superior inter-layer bonding eliminates contaminant unloading and channeling
  • FDA grade polypropylene (DOE only) designed to conform to ANSI/NSF42 & NSF61 standards
  • Narrow range fiber size optimizes consistency of filtration performance
  • Polypropylene construction provides broad chemical compatibility for a variety of applications
  • All materials of construction are FDA listed as acceptable for potable and edible liquid contact according to CFR Title 21
  • Single component construction simplifies compatibility options and provides easy disposal
Applications: Photographic chemical filtration, DI water filtration, plating solution filtration, R.O. pre-filtration, membrane pre-filtration, organic solvent filtration, oil field fluid filtration, bleach filtration, potable water filtration, chemical processing fluid filtration.
Filtration value: 5 µm
Filter structure: Melt blown/spunbonded
Flow: 5GPM per 10" cartridge
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty