Would you like to log in again?
Fulflo® Filter Bags
Supplier: Parker Hannifin
Fulflo®
G2P5-Q
76248-842CS 49.48 USD InStock
76248-842
Fulflo® Filter Bags
Filters Bag Filter Systems
Fulflo® Filter Bags are ideal for virtually any process liquid filtration application requiring the removal of solids. Filter bags are widely used in industrial, inks, paints and coatings and food & beverage industries. Parker’s Fulflo filter bags are manufactured and tested under the strictest quality control standards to assure consistent performance. Parker’s Fulflo filter bags perform at high flow rates and viscosities to 10000 cps or higher.
  • Standard filter bags fit Fulfo bag filter vessels and most major competitive models
  • • The 'C' style Fulflo filter bag features a polypropylene Quik-Seal ring which effectively seals the bag into standard Parker bag vessels
  • • The 'G' style Fulflo® filter bag features a carbon steel snap ring for positive sealing in competitive vessels
  • • Fulflo Quik-Seal™ option is available for all 'G' style Fulflo filter bag media
  • • Available in a range of filter bag media to include polypropylene & polyester felt filter bags, polyester multi-filament filter bags, nylon mono-filament filter bags, and nomex felt filter bags
  • • Felt bags come standard with glazed surface treatment to effectively control migration of fibers into the filtered product
  • • Polypropylene felt (P) bags are suitable for incidental food contact per CFR Title 21
Standard Fulflo Filter Bags are available in 1 to 800 μm particle retention ratings.
Applications: Solvent filtration, bulk chemical filtration, filtration of coatings, coolant filtration, petroleum oil filtration, ink filtration, paint filtration, filtration of adhesives, resin filtration, pre-filters for finer cartridges, filtration for parts washing systems, water filtration.
Seal type: Top sealing plastic ring
Package type: 35 per carton
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty