Would you like to log in again?
Vista® Basic Large Volume Infusion Pump
Supplier: B. Braun
Vista® Basic
637-202
75996-452CS 6717.63 USD InStock
75996-452
Vista® Basic Large Volume Infusion Pump
Pumps Syringe Pumps
The Vista basic was designed to ensure simplicity in all aspects of its use: from set installation, to programming, to special mode use and alarm correction. Based on a design that has sold over 100000 units worldwide, the Vista basic provides high reliability packaged in one of the easiest pumps to use.
  • Robust design of mechanical components ensures maximum durability
  • One-year standard warranty and multiple warranty options are available to meet your future service needs
  • Two-year preventive maintenance schedule helps save and money from typical annual inspection requirements
  • Battery life of approximately 3 years before replacement is required
  • Sets not made with DEHP protect patients, needle-free sets protect both the patient and the clinician
  • Helpful prompts walk the user through the programming process
  • Easy to use, teach and learn, perfect for homecare settings
As a workhorse in the outpatient market, it offers a rugged and reliable design, all relevant therapies such as continuous, piggyback and program mode, and a selection of cost-effective, straight line gravity sets.
Latex information: Not made with natural rubber
Total shelf life: 35 months
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty