Would you like to log in again?
Lighthouse Locking Wall Mount Unit for Sharps Containers
Supplier: Unimed
Lighthouse
SWMU000609
76716-836BX 471.48 USD InStock
76716-836
Lighthouse Locking Wall Mount Unit for Sharps Containers
Bins Sharps Containers
Secures sharps containers in a sturdy locked enclosure.
  • All cabinets are conveniently keyed alike
  • Mounting hardware included
  • Holds Kendall/Covidien 5 quart sharps container with counter balanced lid
  • Made from sturdy ABS plastic for extra protection of the sharps container and long-lasting use
Helps maintain OSHA compliance by keeping used sharps away from patients and personnel, and preventing tipping and spilling of contents. Includes a five quart locking wall mount unit, and a five quart Sharpstar transparent red sharps container.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty