Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triethyl phosphate 99%

Supplier: Ambeed
Danger

A119652-500G A119652-25G A119652-100G
76781-654EA 6.18 USD InStock
76781-654 76781-656 77566-214
Triethyl phosphate 99%
Triethyl phosphate
Caution: For research use only.


Formula: C₆H₁₅O₄P
MW: 182.16 g/mol
MDL Number: MFCD00009077
CAS Number: 78-40-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty