Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Terephthalic acid 95%

Supplier: Ambeed
Warning

A125601-500G A125601-1000G A125601-25G A125601-100G
76781-980EA 19.57 USD InStock
76781-980 76781-982 76781-984 76781-986
Terephthalic acid 95%
Terephthalic acid
Caution: for Research Use Only


Formula: C₈H₆O₄
MW: 166.13 g/mol
MDL Number: MFCD00002558
CAS Number: 100-21-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty