Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Strontium acetate hemihydrate 99%

Supplier: Ambeed
Warning
Synonyms: Strontium Acetate Hemihydrate

A173223-25G A173223-100G A173223-500G
76784-300EA 6.18 USD InStock
76784-300 76784-304 76784-302
Strontium acetate hemihydrate 99%
Strontium acetate hemihydrate
Caution: for Research Use Only


Formula: C₈H₁₄O₉Sr₂
MW: 429,43 g/mol
MDL Number: MFCD00198064
CAS Number: 14692-29-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty