Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trimethylsulfonium iodide 98%

Supplier: Ambeed
Warning

A261511-25G A261511-100G A261511-5G
76787-006EA 58.71 USD InStock
76787-006 76787-008 76787-010
Trimethylsulfonium iodide 98%
Trimethylsulphonium iodide
Caution: for Research Use Only


Formula: C₃H₉IS
MW: 204.07 g/mol
MDL Number: MFCD00011632
CAS Number: 2181-42-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty