Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-3-methyl-L-phenylalanine 98%

Supplier: Ambeed
Warning
Synonyms: Boc-Phe(3-Me)-OH

A270074-250MG A270074-1G A270074-5G
76787-178EA 127.72 USD InStock
76787-178 76787-174 76787-176
N-Boc-3-methyl-L-phenylalanine 98%
N-Boc-3-methyl-L-phenylalanine
Caution: for Research Use Only


Formula: C₁₅H₂₁NO₄
MW: 279.336 g/mol
MDL Number: MFCD01862944
CAS Number: 114873-06-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty