Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetraethylammonium bromide 97%

Supplier: Ambeed
Warning
Synonyms: Tetrylammonium bromide

A272078-500G A272078-100G A272078-1000G A272078-25G
76787-208EA 5.15 USD InStock
76787-208 76787-204 76787-206 76787-210
Tetraethylammonium bromide 97%
Tetraethylammonium bromide
Caution: for Research Use Only


Formula: C₈H₂₀BrN
MW: 210.16 g/mol
MDL Number: MFCD00011825
CAS Number: 71-91-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty