Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Allyl-4,6-dibenzoylresorcinol 98%

Supplier: Ambeed
Warning
Synonyms: (5-Allyl-4,6-dihydroxy-1,3-phenylene)bis(phenylmethanone)

A537070-5G A537070-1G
76790-202EA 15.45 USD InStock
76790-202 76790-204
2-Allyl-4,6-dibenzoylresorcinol 98%
2-Allyl-4,6-dibenzoylresorcinol
Caution: for Research Use Only


Formula: C₂₃H₁₈O₄
MW: 358.40 g/mol
MDL Number: MFCD02094038
CAS Number: 102593-74-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty