Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vinylferrocene 97%

Supplier: Ambeed
Danger

A578954-250MG A578954-1G A578954-5G A578954-25G A578954-10G
76790-400EA 6.18 USD InStock
76790-400 76790-398 76790-396 76790-404 76790-402
Vinylferrocene 97%
Vinylferrocene
Caution: for Research Use Only


Formula: C₁₂H₁₂Fe
MDL Number: MFCD00001434
UN: 1325
ADR: 4.1,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty