Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tartronic acid 98%

Supplier: Ambeed
Warning
Synonyms: 2-Hydroxymalonic acid

A591197-5G A591197-250MG A591197-1G A591197-10G A591197-100MG
76790-498EA 74.16 USD InStock
76790-498 76790-496 76790-494 76790-492 76790-490
Tartronic acid 98%
Tartronic acid
Caution: for Research Use Only


Formula: C₃H₄O₅
MW: 120.06 g/mol
MDL Number: MFCD00004237
CAS Number: 80-69-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty