Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thymolphthalein 96%

Supplier: Ambeed
Warning
Synonyms: 3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one

A746205-5G A746205-100G
76791-278EA 33.99 USD InStock
76791-278 76791-280
Thymolphthalein 96%
Thymolphthaleine
Caution: for Research Use Only


Formula: C₂₈H₃₀O₄
MW: 430.54 g/mol
MDL Number: MFCD00005909
CAS Number: 125-20-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty