Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tris(3-chlorophenyl)phosphine 98%

Supplier: Ambeed
Warning
Synonyms: Tris(m-chlorophenyl)phosphineTri(m-chlorophenyl)phosphine

A808239-250MG A808239-1G
76791-652EA 24.72 USD InStock
76791-652 76791-654
Tris(3-chlorophenyl)phosphine 98%
Tris(3-chlorophenyl)phosphine
Caution: for Research Use Only


Formula: C₁₈H₁₂Cl₃P
MW: 365.63 g/mol
MDL Number: MFCD00013632
CAS Number: 29949-85-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty