Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-Lapachone 98%

Supplier: Ambeed
Warning
Synonyms: 2,2-Dimethyl-3,4-dihydro-2H-benzo[h]chromene-5,6-dione

A951815-5MG A951815-10MG A951815-1MG
76792-194EA 12.36 USD InStock
76792-194 76792-196 76792-192
β-Lapachone 98%
beta-Lapachone
Caution: for Research Use Only


Formula: C₁₅H₁₄O₃
MW: 242.274 g/mol
MDL Number: MFCD01712233
CAS Number: 4707-32-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty