Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Accucold® Ice and Water Dispensers
Supplier: Felix Storch Inc
Accucold®
AIWD282FLTR AIWD282 AIWD450FLTR AIWD160FLTR AIWD160 AIWD450
76748-640EA 13910 USD InStock
76748-640 76748-642 76748-632 76748-634 76748-636 76748-638
Accucold® Ice and Water Dispensers
Water Dispensers
Commercial countertop ice and water dispensers, ideal for waiting areas, break rooms, or similar settings requiring an attractive and efficient water and ice dispenser.
  • Dispenses ice, water, or ice+water to meet all of your user needs
  • Stainless steel construction provides a professional, sleek look
  • UV lighting in the dispenser for improved sanitation
  • Ice shape: Nugget
  • Electrical data: 115 V AC, 60 Hz
Experience the ultimate in convenience with the commercially approved Accucold countertop water and ice dispenser.
Three dispenser functions - ice, water, or ice+water - provide expanded functionality, while the illuminated display ensures easy access and control of the unit.
Adjustable legs offer flexible height options, while the stainless steel finish gives a sleek, professional look. The removable tray and sealed back allow easy cleaning. These units must be used with a drain and should be placed in a space that allows draining below the unit. It is top vented.
Certifications: ETL-S listed to NSF-12 commercial standards. UL, ULC, ETL-S.
Delivery information: They are delivered with 100% guarantee, 1 year-parts and labor and 5 year compressor warranty. The 76748-642, 76748-638 and 76748-634 are supplied with a filter kit.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty