Would you like to log in again?
Eisco® Economy Brass Hooked Weights, Set of 9
Supplier: EISCO SCIENTIFIC LLC
Eisco®
PH0037ECO
470344-054EA 130.85 USD InStock
470344-054
Eisco® Economy Brass Hooked Weights, Set of 9
Weights Calibration Weights
Set of nine brass hooked weights ranging from 10 to 1000 g, perfect for classroom or lab experiments. Supplied in a wooden block for easy storage.
  • 10 to 1000 g brass weights in a wooden block
  • 7.75×3.75×3" wooden block for storage
As this is the economy set, the smaller seven weights are precisely machined, hooked, and made of brass. The two largest weights are a painted brass. All units are inscribed with their specific weight in grams. Wooden block included for storage. The set includes: (1) 1000 g , (1) 500 g, (2) 200 g, (1) 100 g, (1) 50 g, (2) 20 g, (1) 10 g.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty