Would you like to log in again?
Eisco® Slotted Mass Set with Hanger, Stainless Steel, 9 Weights, 500 g
Supplier: EISCO SCIENTIFIC LLC
Eisco®
PH0258NSS
470345-566EA 44.8 USD InStock
470345-566
Eisco® Slotted Mass Set with Hanger, Stainless Steel, 9 Weights, 500 g
Weights Calibration Weights
Set includes nine weights totaling 500 g, made of stainless steel for durability. Set inclues a hanger for suspending.
  • Stainless steel hanger with hook end for suspending masses in force experiments
  • Masses can be used for studying pressure, strength, or torque in the classroom or lab
  • Masses can be used for calibrating scales and balances
Eisco Labs stainless steel slotted masses set can be used with hanging balances as well as for calibration and studying pressure, strength, or torque in the classroom or lab. Unit includes nine weights, and one hanger, each weighing 50 g. Total weight of set is 500 g.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty