Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hexafluorobenzene 97%

Supplier: Ambeed
Danger
Synonyms: Perfluorobenzene

A123811-5G A123811-25G
76797-552EA 31.93 USD InStock
76797-552 76797-554
Hexafluorobenzene 97%
Hexafluorobenzene
Caution: for Research Use Only


Formula: C₆F₆
MW: 186.06 g/mol
MDL Number: MFCD00000288
CAS Number: 392-56-3
UN: 1993
ADR: 3,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty