Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isophthalic acid 98%

Supplier: Ambeed
Warning

A126871-10G A126871-1000G A126871-100G A126871-500G A126871-25G
76797-694EA 20.6 USD InStock
76797-694 76797-698 76797-696 76797-702 76797-700
Isophthalic acid 98%
Isophthalic acid
Caution: for Research Use Only


Formula: C₈H₆O₄
MW: 166.13 g/mol
MDL Number: MFCD00002516
CAS Number: 121-91-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty