Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diphenyl(o-tolyl)phosphine 97%

Supplier: Ambeed
Warning

A203169-1G A203169-5G A203169-25G A203169-250MG
76800-434EA 63.86 USD InStock
76800-434 76800-428 76800-432 76800-430
Diphenyl(o-tolyl)phosphine 97%
Diphenyl(o-tolyl)phosphine
Caution: for Research Use Only


Formula: C₁₉H₁₇P
MW: 276.32 g/mol
MDL Number: MFCD00008515
CAS Number: 5931-53-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty