Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DL-Phenylalaninol 97%

Supplier: Ambeed
Danger
Synonyms: DL-2-Amino-3-phenyl-1-propanol2-Amino-3-phenyl-1-propanol2-Amino-3-phenylpropan-1-ol

A236222-100G
76801-216EA 214.24 USD InStock
76801-216
DL-Phenylalaninol 97%
DL-Phenylalaninol
Caution: for Research Use Only


Formula: C₉H₁₃NO
MW: 151.21 g/mol
MDL Number: MFCD00066689
CAS Number: 16088-07-6
UN: 3259
ADR: 8,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty