Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium chloride hexahydrate 98%

Supplier: Ambeed
Warning

A252423-25G A252423-500G A252423-100G
76801-586EA 5.15 USD InStock
76801-586 76801-588 76801-584
Magnesium chloride hexahydrate 98%
Magnesium chloride hexahydrate
Caution: for Research Use Only


Formula: MgCl₂·6H₂O
MW: 203.3 g/mol
MDL Number: MFCD00149781
CAS Number: 7791-18-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty