Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Benzyl-D-glutamate 97%

Supplier: Ambeed
Warning
Synonyms: D-Glutamic Acid 1-Benzyl EsterH-D-Glu-OBzl

A270779-5G A270779-1G A270779-25G
76801-976EA 15.45 USD InStock
76801-976 76801-978 76801-980
1-Benzyl-D-glutamate 97%
1-Benzyl-D-glutamate
Caution: for Research Use Only


Formula: C₁₂H₁₅NO₄
MW: 237.26 g/mol
MDL Number: MFCD00237065
CAS Number: 79338-14-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty