Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Leucine-p-nitroanilide 98%

Supplier: Ambeed
Warning
Synonyms: H-Leu-pNA(S)-2-Amino-4-methyl-N-(4-nitrophenyl)pentanamide

A283905-250MG
76802-266EA 19.57 USD InStock
76802-266
L-Leucine-p-nitroanilide 98%
L-Leucine-p-nitroanilide
Caution: for Research Use Only


Formula: C₁₂H₁₇N₃O₃
MW: 251.286 g/mol
MDL Number: MFCD00063681
CAS Number: 4178-93-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty