Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl terephthalate 98%

Supplier: Ambeed
Warning
Synonyms: Terephthalic acid dimethyl ester

A303851-100G A303851-1000G A303851-500G A303851-25G
76802-572EA 17.51 USD InStock
76802-572 76802-578 76802-576 76802-574
Dimethyl terephthalate 98%
Dimethyl terephthalate
Caution: for Research Use Only


Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
MDL Number: MFCD00008440
CAS Number: 120-61-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty