Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

H-D-Asp-OMe 95%

Supplier: Ambeed
Warning
Synonyms: D-Aspartic acid 1-methyl ester(R)-3-Amino-4-methoxy-4-oxobutanoic acid

A374003-10G A374003-100G
76803-644EA 69.01 USD InStock
76803-644 76803-642
H-D-Asp-OMe 95%
H-D-Asp-OMe
Caution: for Research Use Only


Formula: C₅H₉NO₄
MW: 147.13 g/mol
MDL Number: MFCD03424227
CAS Number: 65414-78-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty