Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl diselenide 95%

Supplier: Ambeed
Danger
Synonyms: Dimethyldiselenide

A395532-25G A395532-1G
77594-210EA 18 USD InStock
77594-210 76803-896
Dimethyl diselenide 95%
Dimethyl diselenide
Caution: for Research Use Only


Formula: C₂H₆Se₂
MW: 187.99 g/mol
MDL Number: MFCD00008254
CAS Number: 7101-31-7
UN: 1992
ADR: 3,6.1,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty