Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methoxyacetic acid 98%

Supplier: Ambeed
Danger
Synonyms: 2-Methoxyacetic acid

A424777-100G A424777-25G A424777-500G
76804-182EA 51.5 USD InStock
76804-182 76804-180 76804-178
Methoxyacetic acid 98%
Methoxyacetic acid
Caution: for Research Use Only


Formula: C₃H₆O₃
MW: 90.08 g/mol
MDL Number: MFCD00004308
CAS Number: 625-45-6
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty