Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gold tribromide 98%

Supplier: Ambeed
Danger
Synonyms: Gold (III) bromideGold(III) bromide

A490595-250MG A490595-1G
76804-876EA 57.68 USD InStock
76804-876 76804-874
Gold tribromide 98%
Gold tribromide
Caution: for Research Use Only


Formula: AuBr₃
MW: 436.68 g/mol
MDL Number: MFCD00014171
CAS Number: 10294-28-7
UN: 3260
ADR: 8,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty