Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gold trihydroxide 98%

Supplier: Ambeed
Warning
Synonyms: Gold(iii)hydroxide

A522387-1G A522387-100MG A522387-250MG
76805-180EA 176.13 USD InStock
76805-180 76805-182 76805-178
Gold trihydroxide 98%
Gold trihydroxide
Caution: for Research Use Only


Formula: H₃AuO₃
MW: 247.99 g/mol
MDL Number: MFCD00046173
CAS Number: 1303-52-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty