Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethoxyacetic acid 98%

Supplier: Ambeed
Danger
Synonyms: 2-Ethoxyacetic acid

A565827-1G A565827-5G A565827-10G A565827-25G
76805-668EA 6.18 USD InStock
76805-668 76805-662 76805-664 76805-666
Ethoxyacetic acid 98%
Ethoxyacetic acid
Caution: for Research Use Only


Formula: C₄H₈O₃
MW: 104.11 g/mol
MDL Number: MFCD00004310
CAS Number: 627-03-2
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty