Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hexanophenone 98%

Supplier: Ambeed
Warning

A662968-100G A662968-25G A662968-5G
76806-472EA 9.27 USD InStock
76806-472 76806-470 76806-468
Hexanophenone 98%
Hexanophenone
Caution: for Research Use Only


Formula: C₁₂H₁₆O
MW: 176.26 g/mol
MDL Number: MFCD00009512
CAS Number: 942-92-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty