Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ferrocene 98%

Supplier: Ambeed
Danger

A672285-25G
76806-564EA 5.15 USD InStock
76806-564
Ferrocene 98%
Ferrocene
Caution: for Research Use Only


Formula: C₁₀H₁₀Fe
MW: 186.04 g/mol
MDL Number: MFCD00001427
CAS Number: 102-54-5
UN: 1325
ADR: 4.1,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty