Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Indoleacrylic acid 98%

Supplier: Ambeed
Warning
Synonyms: 3-(3-Indolyl)acrylic AcidIAA3-(1H-Indol-3-yl)acrylic acid

A692274-5G A692274-1G
76806-658EA 45.32 USD InStock
76806-658 76806-656
3-Indoleacrylic acid 98%
3-Indoleacrylic acid
Caution: for Research Use Only


Formula: C₁₁H₉NO₂
MW: 187.20 g/mol
MDL Number: MFCD00005633
CAS Number: 1204-06-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty