Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fmoc-4-(boc-aminomethyl)-L-phenylalanine 98%

Supplier: Ambeed
Warning
Synonyms: Fmoc-4-(Boc-aminomethyl)-L-phenylalanine

A745611-100MG A745611-250MG
76807-120EA 84.46 USD InStock
76807-120 76807-118
Fmoc-4-(boc-aminomethyl)-L-phenylalanine 98%
Fmoc-4-(boc-aminomethyl)-L-phenylalanine
Caution: for Research Use Only


Formula: C₃₀H₃₂N₂O₆
MW: 516.6 g/mol
MDL Number: MFCD01317015
CAS Number: 204715-91-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty