Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium acetate dihydrate 99%

Supplier: Ambeed
Warning
Synonyms: Acetic acid lithium salt

A765248-25G A765248-1000G
76807-276EA 6.18 USD InStock
76807-276 76807-274
Lithium acetate dihydrate 99%
Lithium acetate dihydrate
Caution: for Research Use Only


Formula: H₃CCOOLi·2H₂O
MW: 102.02 g/mol
MDL Number: MFCD00066949
CAS Number: 6108-17-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty