Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eisco® Stainless Steel Slotted Mass Hanger, 100 g

Supplier: EISCO SCIENTIFIC LLC
Eisco®
PHSSH100
470345-260EA 9.19 USD InStock
470345-260
Eisco® Stainless Steel Slotted Mass Hanger, 100 g
Weights Calibration Weights
Supports stainless steel masses up to 100 g. Suitable for use in physics experiments.

  • Stainless steel hanger weighing 100 g
  • Supports stainless steel slotted masses up to 100 g
  • Overall height measures 6.5"
  • Length of mass placement section (bottom of hook to top of base) measures 4.5"
  • Base diameter measure 1.57"

This premium quality, durable stainless steel slotted masses hanger weighs 100 g and supports stainless steel slotted masses up to 100 g (item numbers (PHMSSS01 through PHMSSS07). Overall height from top of hook to bottom of base measures 6.5" tall, 4.5" of which accommodates mass placement. A 1.57" diameter base provides added stability.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty