Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eisco® Zinc Casted Slotted Masses, Set of 9 Weights, 180 g Total

Supplier: EISCO SCIENTIFIC LLC
Eisco®
PHZNW200SET
470345-838EA 14.9 USD InStock
470345-838
Eisco® Zinc Casted Slotted Masses, Set of 9 Weights, 180 g Total
Weights Calibration Weights
Set of 9 zinc-casted slotted masses with one hanger each of 20 g, total weight of 180 g. For use in physics experiments and demonstrations.

  • 9 Weights
  • 20 g per weight, 200 g total (9 weight set)
  • Hanger for weights included
  • Base diameter: 1.38"
  • Weight diameter: 1.38"

This set of 9 weights, 20 g each (total of 200 g) are easily placed on and taken off the hanger (included), making them very user friendly for experiments involving hanging weights on rulers or springs.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty